missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid, 97%, Thermo Scientific™

105.00 € - 282.00 €

Chemical Identifiers

CAS 212578-38-6
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD11506351
InChI Key SSVIRLKDUUXSTR-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-2-carboxylic acid, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid, 2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl, 4-methyl-2h,3h-benzo e 1,4-oxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazole-2-carboxylic acid
PubChem CID 10845449
IUPAC Name 4-methyl-2,3-dihydro-1,4-benzoxazine-2-carboxylic acid
SMILES CN1CC(OC2=CC=CC=C21)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10537274
SDS View Documents
Thermo Scientific Maybridge
CC29801CB
250 mg Amber Glass Bottle
105.00 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10693615
SDS View Documents
Thermo Scientific Maybridge
CC29801DA
1 g Amber Glass Bottle
282.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.