missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrophenyl isocyanate, 96%, Thermo Scientific™

Product Code. 11303306
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13471-69-7
Molecular Formula C8H6N2O3
Molecular Weight (g/mol) 178.147
MDL Number MFCD00014698
InChI Key OIORBBLUSMONPW-UHFFFAOYSA-N
PubChem CID 73137
IUPAC Name 4-isocyanato-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)N=C=O)[N+](=O)[O-]

Specifications

Melting Point 48°C to 54°C
Boiling Point 113°C to 118°C (3 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Quantity 1 g
UN Number UN2811
Beilstein 2724128
Synonym 4-methyl-3-nitrophenyl isocyanate, 2-nitro-4-isocyanatotoluene, 4-methyl-3-nitrophenylisocyanate, benzene, 4-isocyanato-1-methyl-2-nitro, benzene,4-isocyanato-1-methyl-2-nitro, 4-methyl-3-nitrobenzenisocyanate, acmc-20amdt
Formula Weight 178.15
Percent Purity 96%
Chemical Name or Material 4-Methyl-3-nitrophenyl isocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.