missing translation for 'onlineSavingsMsg'
Learn More
4-Methylpyridine-3-boronic acid, 90%, Thermo Scientific Chemicals Product Code.: 10575670

4-Methylpyridine-3-boronic acid, 90%, Thermo Scientific Chemicals

Product Code. 10575670
1 g, Glass bottle
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10575670

Brand: Thermo Scientific Acros 430270010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 148546-82-1
CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H8BNO2
MDL Number MFCD03788244
Quantity 1 g
Synonym 4-methylpyridine-3-boronic acid, 4-methylpyridin-3-yl boronic acid, 4-methylpyridin-3-yl-3-boronic acid, 4-methyl-3-pyridyl boronic acid, 3-borono-4-picoline, boronic acid, 4-methyl-3-pyridinyl, 4-methyl-3-pyridinyl boronic acid, 4-picoline-3-boronic acid, 4-picoline-5-boronic acid, 4-methyl-3-pyridinyl-boronic acid
InChI Key ASXFMIDIRZPCGK-UHFFFAOYSA-N
SMILES B(C1=C(C=CN=C1)C)(O)O
IUPAC Name (4-methylpyridin-3-yl)boronic acid
Molecular Weight (g/mol) 136.95
PubChem CID 2762711
Formula Weight 136.95
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 4-Methylpyridine-3-boronic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number339693 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number430270010
compliance-icons
Product Identifier
  • 4-Methylpyridine-3-boronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methylpyridine-3-boronic acid, 90%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.