missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylbenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 11453917
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11453917

Brand: Thermo Scientific Alfa Aesar A18354.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146949-07-7
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.70
MDL Number MFCD00041508
InChI Key LEFGAGRZHLNPLS-UHFFFAOYSA-N
Synonym 4-n-propylbenzenesulfonyl chloride, 4-propylbenzene-1-sulfonyl chloride, 4-propyl-benzenesulfonylchloride, 4-propyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-propyl, p-propylbenzenesulfonyl chloride, 4-n-propylbenzenesulfonylchloride, acmc-209cxt, 4-propylphenylsulfonyl chloride, 4-propylphenylsulphonyl chloride
PubChem CID 2737222
IUPAC Name 4-propylbenzenesulfonyl chloride
SMILES CCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 20°C to 22°C
Density 1.223
Boiling Point 275°C to 276°C
Refractive Index 1.5375
Quantity 5 g
UN Number UN3265
Beilstein 5511216
Sensitivity Moisture sensitive
Formula Weight 218.7
Percent Purity 95%
Chemical Name or Material 4-n-Propylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.