missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nonanone, 97+%, Thermo Scientific™
32.90 € - 367.00 €
Chemical Identifiers
CAS | 4485-09-0 |
---|---|
Molecular Formula | C9H18O |
Molecular Weight (g/mol) | 142.24 |
MDL Number | MFCD00027286 |
InChI Key | TYBCSQFBSWACAA-UHFFFAOYSA-N |
Synonym | 4-nonanone, amyl propyl ketone, propyl amyl ketone, pentyl propyl ketone, amylpropylketone, acmc-1ailr, n-pentyl n-propyl ketone |
PubChem CID | 78236 |
IUPAC Name | nonan-4-one |
SMILES | CCCCCC(=O)CCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11477317
|
Thermo Scientific Alfa Aesar
A15050.14 |
25 g | N/A | N/A | N/A | |||||
11487317
|
Thermo Scientific Alfa Aesar
A15050.22 |
100 g | N/A | N/A | N/A | |||||
11497317
|
Thermo Scientific Alfa Aesar
A15050.36 |
500 g | N/A | N/A | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
4485-09-0 | |
142.24 | |
TYBCSQFBSWACAA-UHFFFAOYSA-N | |
78236 | |
CCCCCC(=O)CCC |
C9H18O | |
MFCD00027286 | |
4-nonanone, amyl propyl ketone, propyl amyl ketone, pentyl propyl ketone, amylpropylketone, acmc-1ailr, n-pentyl n-propyl ketone | |
nonan-4-one |
Specifications
4485-09-0 | |
187°C | |
MFCD00027286 | |
TYBCSQFBSWACAA-UHFFFAOYSA-N | |
nonan-4-one | |
78236 | |
≥97% |
0.819 | |
C9H18O | |
4-nonanone, amyl propyl ketone, propyl amyl ketone, pentyl propyl ketone, amylpropylketone, acmc-1ailr, n-pentyl n-propyl ketone | |
CCCCCC(=O)CCC | |
142.24 | |
142.24 | |
4-Nonanone |
Safety and Handling
missing translation for 'einecsNumber' : 224-770-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Nonanone, 97+%, Thermo Scientific™