missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Carboxyphenyl)-1,2,4-triazole, 97%, Thermo Scientific™

Product Code. 15552303 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15552303

Brand: Thermo Scientific Acros 449960050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 157069-48-2
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.17
InChI Key QCNDLFQKKMPZKH-UHFFFAOYSA-N
Synonym 4-4h-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4 triazol-4-yl-benzoic acid, benzoic acid, 4-4h-1,2,4-triazol-4-yl, 4-4-carboxyphenyl-1,2,4-triazole, 4-4-carboxyphenyl-4h-1,2,4-triazole, 4-4h-1,2,4-triazol-4-yl-benzoic acid
PubChem CID 3163581
IUPAC Name 4-(1,2,4-triazol-4-yl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)N2C=NN=C2

Specifications

Melting Point 330.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 4-(p-Carboxyphenyl)-1,2,4-triazole
compliance-icons
Product Identifier
  • 4-(p-Carboxyphenyl)-1,2,4-triazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.