missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

40.60 € - 133.00 €

Chemical Identifiers

CAS 133115-72-7
Molecular Formula C7H8ClF3N2O
Molecular Weight (g/mol) 228.60
MDL Number MFCD00053033
InChI Key KQXZVSQCMVKMBK-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenylhydrazine hydrochloride, 4-trifluoromethoxy phenyl hydrazine hydrochloride, 4-trifluoromethoxyphenylhydrazine hydrochloride, 4-trifluoromethoxy phenylhydrazine hcl, 4-trifluoromethoxyphenyl-hydrazine hcl, 1-4-trifluoromethoxy phenyl hydrazine hydrochloride, p-trifluoromethoxy phenylhydrazine hydrochloride, hydrazine, 4-trifluoromethoxy phenyl-, monohydrochloride
PubChem CID 2777328
IUPAC Name [4-(trifluoromethoxy)phenyl]hydrazine;hydrochloride
SMILES Cl.NNC1=CC=C(OC(F)(F)F)C=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10586874
SDS View Documents
Thermo Scientific Acros
397040010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10041754
SDS View Documents
Thermo Scientific Acros
397040050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.