Learn More
B Braun™ Needle-free IV Injection Port Valve
Click to view available options
Unit Size:
Pack of 100
Description
- Lipid-resistant material
- Flow rates up to 200mL/min.
- 0.05mL positive displacement
- Compatible with Luer Lock and Luer Slip, valve opens and closes automatically with each Luer taper
- Not made with DEHP, PVC, or natural rubber latex
- Simplifies handling
- Helps prevent bloodstream infections
- Helps reduce catheter occlusions
Specifications
Specifications
| Max. Pressure | 300 psi |