missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(trifluoromethyl)benzimidazole, 97+%, Thermo Scientific Chemicals

64.82 € - 230.00 €

Chemical Identifiers

CAS 3671-66-7
Molecular Formula C8H6F3N3
Molecular Weight (g/mol) 201.152
MDL Number MFCD01632182
InChI Key CKEKFQLHCAZGSP-UHFFFAOYSA-N
Synonym 5-amino-2-trifluoromethyl benzimidazole, 1h-benzimidazol-5-amine, 2-trifluoromethyl, 2-trifluoromethyl-1h-benzo d imidazol-5-amine, benzimidazole, 5-amino-2-trifluoromethyl, 2-trifluoromethyl-1h-benzimidazol-5-amine, 2-trifluoromethyl-1h-1,3-benzodiazol-5-amine, 5-amino-2-trifluoromethylbenzimidazole, 2-trifluoromethyl benzimidazole-5-ylamine, 2-trifluoromethyl-1h-benzimidazol-6-amine, 2-trifluoromethyl-3h-benzoimidazol-5-ylamineShow MoreShow Less
PubChem CID 19345
IUPAC Name 2-(trifluoromethyl)-3H-benzimidazol-5-amine
SMILES C1=CC2=C(C=C1N)NC(=N2)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11409807
SDS View Documents
Thermo Scientific Alfa Aesar
B22770.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11419807
SDS View Documents
Thermo Scientific Alfa Aesar
B22770.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'rtecsNumber' : DD6205700

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Amino-2-(trifluoromethyl)benzimidazole, 97+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.