missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiazole, 97%

Product Code. 11302807
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11302807

Brand: Thermo Scientific Alfa Aesar L15879.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3581-89-3 | C4H5NS | 99.151 g/mol

5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to yeild mobile, room-temperature ionic liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to yeild mobile, room-temperature ionic liquids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3581-89-3
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.151
MDL Number MFCD00005335
InChI Key RLYUNPNLXMSXAX-UHFFFAOYSA-N
Synonym 5-methylthiazole, thiazole, 5-methyl, 5-methyl-thiazole, 5-methyl thiazole, zlchem 1239, acmc-1cjhk, #, ksc222m5d
PubChem CID 137980
IUPAC Name 5-methyl-1,3-thiazole
SMILES CC1=CN=CS1

Specifications

Density 1.12
Boiling Point 141°C to 142°C
Flash Point 42°C (107°F)
Odor Stench
Refractive Index 1.528
Quantity 5 g
UN Number UN1993
Beilstein 104674
Solubility Information Insoluble in water.
Formula Weight 99.16
Percent Purity 97%
Chemical Name or Material 5-Methylthiazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.