Learn More
Greiner Bio-One 5 mL Sterile Serological Pipets
Drip-free pipetting design with high optical clarity and maximum accuracy.
142.00 € - 283.00 €
Specifications
Material | Polystyrene |
---|---|
Sterility | Sterile |
Graduated | Yes |
Graduations | 1/10 |
Volume (Metric) | 5 mL |
Product Code | Brand | Packaging Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Price | Quantity & Availability | |||||
13002013
|
Greiner Bio-One
606160 |
Individual Plastic Wrap |
142.00 €
Pack of 200 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||||
10018810
|
Greiner Bio-One
606107 |
Bulk |
283.00 €
Pack of 500 |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Description
- High-grade polystyrene
- Clear, easily legible graduation
- Colored vertical stripe for ease in reading the volume
- With synthetic fibrous filter material for protection against
Specifications
Polystyrene | |
Yes | |
5 mL |
Sterile | |
1/10 | |
Serological Pipet |