Learn More
Greiner Bio-One 50 mL Sterile Serological Pipets
GREENER_CHOICE
Drip-free pipetting design with high optical clarity and maximum accuracy.
252.00 €
Specifications
Material Polystyrene
Sterility Sterile
Graduated Yes
Graduations 1/2
Volume (Metric) 50 mL
View More Specs
Products 1
Product Code Brand Packaging Type Price Quantity & Availability  
Product Code Brand Packaging Type Price Quantity & Availability  
10561804
GREENER_CHOICE
View Documents
Greiner Bio-One
768160
Individual Plastic Wrap
252.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
Specifications
Specifications
Polystyrene
Yes
50 mL
Sterile
1/2
Serological Pipet
Videos
SDS
Documents
Documents
Product Certifications