missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Mg, AA 5283, A95283, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Aluminum, Al/Mg, AA 5283, A95283, 50g
Specifications
Specifications
| Chemical Name or Material | Beryllium/Chromium/Nickel/Titanium/Copper/Zinc/Silicon/Iron/Tin/Lead/Manganese/Magnesium |
| CAS | 7429-90-5 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Beryllium 0.014 %/Chromium 0.029 %/Nickel 0.04 %/Titanium 0.048 %/Copper 0.056 %/Zinc 0.061 %/Silicon 0.11 %/Iron 0.14 %/Tin 0.14 %/Lead 0.15 %/Manganese 0.55 %/Magnesium 5.1 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction