missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-2-fluoro-3-methoxybenzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 1 g |
| Synonym | 6-chloro-2-fluoro-3-methoxybenzoicacid, benzoic acid,6-chloro-2-fluoro-3-methoxy, benzoic acid, 6-chloro-2-fluoro-3-methoxy, 6-chloro-2-fluoro-3-methyloxy benzoic acid, 6-chloranyl-2-fluoranyl-3-methoxy-benzoic acid, 6-chloro-2-fluoro-3-methoxybenzenecarboxylic acid |
| Formula Weight | 204.59 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Chloro-2-fluoro-3-methoxybenzoic acid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion