missing translation for 'onlineSavingsMsg'
Learn More

6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester, 97%

Product Code. 11434610
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11434610

Brand: Thermo Scientific Alfa Aesar H50139.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 947191-69-7 | C15H22BNO3 | 275.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947191-69-7
Molecular Formula C15H22BNO3
Molecular Weight (g/mol) 275.155
MDL Number MFCD07781184
InChI Key QEQNYJUPYICBCR-UHFFFAOYSA-N
Synonym 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-cyclopropylmethoxy pyridine-3-boronic acid pinacol ester, 2-cyclopropylmethoxy pyridine-5-boronic acid, pinacol ester, 6-cyclopropylmethoxypyridine-3-boronic acid pinacol ester, 6-cyclopropylmethoxypyridine-3-yl boronic acid pinacol ester, 6-cyclopropylmethoxy pyridin-3-yl boronic acid pinacol ester, 2-cyclopropyl-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 24208797
IUPAC Name 2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC3CC3

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 275.16
Percent Purity 97%
Chemical Name or Material 6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.