missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methylheptanoic acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Methylheptanoic acid is used as a reagent to prepare eponemycin analogues, which is an antibiotic. Further, it is an active component of the scent chemical of the brushtail possum. In addition to this, it is used in the preparation of 6-methyl-heptanoyl chloride.
Solubility
Miscible with chloroform and methanol. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents. Store in a cool place.
6-Methylheptanoic acid is used as a reagent to prepare eponemycin analogues, which is an antibiotic. Further, it is an active component of the scent chemical of the brushtail possum. In addition to this, it is used in the preparation of 6-methyl-heptanoyl chloride.
Solubility
Miscible with chloroform and methanol. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents. Store in a cool place.
Specifikationer
Specifikationer
| Refractive Index | 1.4275 |
| Quantity | 1 g |
| Beilstein | EM>10 |
| Solubility Information | Miscible with chloroform and methanol. Slightly miscible with water. |
| Formula Weight | 144.22 |
| Percent Purity | 95% |
| Chemical Name or Material | 6-Methylheptanoic acid |
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering