missing translation for 'onlineSavingsMsg'
Learn More
7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals Product Code.: 15407528

7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

Product Code. 15407528
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15407528

Brand: Thermo Scientific Alfa Aesar H61032.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Sparingly soluble in water (0.76 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120711-81-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD11603433
InChI Key YKCFDUNYLMTXFC-UHFFFAOYSA-N
Synonym 7-nitro-3,4-dihydro-2h-1,4-benzooxazine, 7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 2,3-dihydro-7-nitro-1,4-benzoxazine, 3,4-dihydro-7-nitro-2h-1,4-benzoxazine, 7-nitro-2,3-dihydro-4h-1,4-benzoxazine, 2h-1,4-benzoxazine, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine, 7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine
PubChem CID 18416151
IUPAC Name 7-nitro-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]

Specifications

Quantity 250 mg
Solubility Information Sparingly soluble in water (0.76g/L at 25°C).
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 7-Nitro-3,4-dihydro-2H-1,4-benzoxazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.