Learn More
Knick Elektronische™ 766 Waterproof Laboratory pH Meter
Product Code. 10518181
Click to view available options
IP Rating:
IP54
Unit Size:
Each
Product Code. 10518181
Brand: Knick Elektronische™ 766
Only null left
Add to basket
Add to basket
The 766 Waterproof Laboratory pH Meter is designed for standard applications in everyday lab routines. It combines practical functionality and easy operation with comprehensive safety functions.
  • Gaincheck automatic device test
  • Trueline calibrated analog recorder output
  • Sensor monitoring with Sensoface icon alerts
  • Automatic calibration with patented Calimatic
  • EMC according to NAMUR
  • Simultaneous pH and temperature display
  • Simple operation
  • Leak-proof membrane keyboard
  • Robust housing
  • Protection category IP 54
  • 3-year warranty
gaincheck, Calimatic, record, recorders, pH, meter
Specifications
Includes Sensoface Icon Alerts, Automatic Calibration, Simultaneous pH and Temperature Display
For Use With (Application) Standard applications in everyday lab routines
IP Rating IP54
Keypad Leak-proof membrane keyboard
Model 766 Laboratory pH Meter
Type Laboratory pH Meter