Learn More
Brady™ 7942 Series Thermal Transfer Printer Ribbon
General purpose wax ribbon for paper or temporary labeling.
Brand:  Brady™ 237558
 View more versions of this product
Product Code. 11533155
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
General purpose, standard performance wax ribbon for paper and temporary labeling.
Specifications
Specifications
Black
Wax
1.6 in.
40 mm
25.4 mm
2011/65/EU and 2015/863
BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer, THT 76 mm Core Printer
984 ft.
300 m
1 in.
Open-core Ribbon