Learn More
Brady™ 7942 Series Thermal Transfer Printer Ribbon
General purpose wax ribbon for paper or temporary labeling.
Brand:  Brady™ 237561
 View more versions of this product
Product Code. 11553155
  • 88.05 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
General purpose, standard performance wax ribbon for paper and temporary labeling.
Specifications
Specifications
Black
Wax
4.3 in.
110 mm
25.4 mm
2011/65/EU and 2015/863
BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer, THT 76 mm Core Printer
984 ft.
300 m
1 in.
Open-core Ribbon