Learn More
Brady™ 7950 Series Thermal Transfer 25.4 mm Core Printer Ribbon, 110 mm Width
Thermal transfer ribbon with high resistance against smearing and chemicals.
Brand: Brady™ 236003
This item is not returnable.
View return policy
Description
Strong environmental resistance against smearing and chemicals.
- Reliable, sharp-edge print quality maximizes bar code readability
Specifications
Black | |
Wax, Resin | |
4.3 in. | |
110 mm | |
25.4 mm | |
2011/65/EU and 2015/863 |
BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer | |
984 ft. | |
300 m | |
1 in. | |
Open-core Ribbon | |
R7950-110X300/O |