Learn More
Brady™ 7961 Series Thermal Transfer Printer Ribbon
Thermal transfer ribbon with excellent scratch and good chemical resistance.
Brand: Brady™ 236015
This item is not returnable.
View return policy
Description
When printed on the appropriate Brady™ label, this ribbon gives excellent scratch and good chemical resistance.
Specifications
Black | |
Resin | |
4.5 in. | |
114 mm | |
25.4 mm | |
2011/65/EU and 2015/863 |
BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer, THT 76 mm Core Printer | |
984 ft. | |
300 m | |
1 in. | |
Open-core Ribbon | |
R7961-114X300/O |