Learn More
Brady™ 7962 Series Thermal Transfer Printer Ribbon
Thermal transfer ribbon with high smear, scratch, solvent, and fuel resistance.
Brand: Brady™ 236023
This item is not returnable.
View return policy
Description
R7962 is a high performance resin ribbon with high smear and scratch resistance and excellent solvent and gas fuel resistance.
Specifications
Black | |
Resin | |
4.3 in. | |
110 mm | |
25.4 mm | |
R7962-110X300/O |
BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer | |
984 ft. | |
300 m | |
1 in. | |
2011/65/EU and 2015/863 |