missing translation for 'onlineSavingsMsg'
Learn More
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™ Product Code.: 15405215

8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Product Code. 15405215
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15405215

Brand: Thermo Scientific Alfa Aesar H35662.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353258-35-2
Molecular Formula C9H6ClF3N2O3
Molecular Weight (g/mol) 282.603
MDL Number MFCD22682820
InChI Key HBDANCBTRVDEBT-UHFFFAOYSA-N
Synonym 8-chloro-6-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid hydrate
PubChem CID 122361169
IUPAC Name 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid;hydrate
SMILES C1=C(C2=NC(=CN2C=C1C(F)(F)F)C(=O)O)Cl.O

Specifications

Melting Point ∽188°C (decomposition)
Quantity 1 g
Formula Weight 264.59 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.