missing translation for 'onlineSavingsMsg'
Learn More

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride, 97%

Product Code. 15400739
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15400739

Brand: Thermo Scientific Alfa Aesar H63825.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54745-74-3 | C6H12ClNO | 149.62 g/mol

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54745-74-3
Molecular Formula C6H12ClNO
Molecular Weight (g/mol) 149.62
MDL Number MFCD17926456,MFCD09800611
InChI Key XADOTNAXKKFKDY-UHFFFAOYNA-N
Synonym 8-oxa-3-azabicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclo 3,2,1 octane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane hcl, 8-oxa-3-azabicyclo 3.2.1 octanehydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane, hydrochloride, 8-oxa-3-aza-bicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclooctane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane-hcl, 8-oxa-3-aza-bicyclo 3.2.1 octane hcl
PubChem CID 21983536
SMILES Cl.C1CC2CNCC1O2

Specifications

Quantity 5 g
IUPAC Name 8-oxa-3-azabicyclo[3.2.1]octane hydrochloride
Formula Weight 149.62
Percent Purity 97%
Chemical Name or Material 8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.