Learn More
Greiner Bio-One 8-well Polystyrene Round Bottom Strip Plates
Free of detectable DNase/RNase, human DNA and pyrogens.
Brand: Greiner Bio-One 767001
This item is not returnable.
View return policy
Description
- 1 x 8 strips
- Non-Sterile
- Round Bottom
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Strip Plate | |
Polystyrene | |
Non-sterile | |
Round Bottom (U-well) |
Medium Binding | |
8 | |
Microlon 200 | |
400 |