Learn More
9,9-Di-n-hexylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals
16.58 € - 141.71 €
Chemical Identifiers
CAS | 203927-98-4 |
---|---|
Molecular Formula | C25H36B2O4 |
Molecular Weight (g/mol) | 422.18 |
MDL Number | MFCD03427279 |
InChI Key | QFWOJNOZWMHNTK-UHFFFAOYSA-N |
Synonym | 9,9-dihexylfluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid, 7-borono-9,9-dihexyl-fluoren-2-yl boronic acid, boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis, acmc-20alvw, 9,9-dihexyl-fluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diboronic acid, 7-borono-9,9-dihexylfluoren-2-yl boronic acid, 2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluoreneShow More |
PubChem CID | 4524285 |
SMILES | CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15492306
|
Thermo Scientific Alfa Aesar
H55817.MD |
250 mg | N/A | ||||||
15462168
|
Thermo Scientific Alfa Aesar
H55817.03 |
1 g |
141.71 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
203927-98-4 | |
422.18 | |
QFWOJNOZWMHNTK-UHFFFAOYSA-N | |
4524285 |
C25H36B2O4 | |
MFCD03427279 | |
9,9-dihexylfluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid, 7-borono-9,9-dihexyl-fluoren-2-yl boronic acid, boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis, acmc-20alvw, 9,9-dihexyl-fluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diboronic acid, 7-borono-9,9-dihexylfluoren-2-yl boronic acid, 2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluoreneShow More | |
CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O |
Specifications
203927-98-4 | |
MFCD03427279 | |
QFWOJNOZWMHNTK-UHFFFAOYSA-N | |
(7-borono-9,9-dihexylfluoren-2-yl)boronic acid | |
4524285 | |
97% | |
9,9-Di-n-hexylfluorene-2,7-diboronic acid |
C25H36B2O4 | |
9,9-dihexylfluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid, 7-borono-9,9-dihexyl-fluoren-2-yl boronic acid, boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis, acmc-20alvw, 9,9-dihexyl-fluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diboronic acid, 7-borono-9,9-dihexylfluoren-2-yl boronic acid, 2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluorene | |
CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O | |
422.18 | |
422.18 | |
Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.