Description
- Comfortable cover spectacle with frosted brow guard to block overhead glare
- Indirect ventilation due to special temple design with large side shields
- Supravision plus coating is scratch-resistant and anti-fog on both sides
Specifications
Specifications
Lens Type | Supravision Plus |
Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
Lens Tint | Clear |
Frame Color | Blue, Clear |
Lens Material | Polycarbonate |
UV Protection | UV400 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Uvex™ 9161 Coated Safety Spectacles >
Spot an opportunity for improvement?Share a Content Correction