Learn More
Eppendorf™ 96-channel Dispensing Heads
For use with epMotion™ 96 Flex in hard-shell case. Requires base device for operation.
Brand: Eppendorf™ 5069000527
7250.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with epMotion™ 96 Flex (not included)
- Intuitive semi-automated handling of 96- and 384-well plates
- Calibrated according to ISO 23783-2 standards
- Includes hard shell case for transport and storage
Specifications
Dispensing Head | |
0.5 to 300 ÎĽL | |
epMotion™ 96 Flex, and epT.I.P.S.™ Motion Reload System | |
4.6 lb. | |
1 Ea. |
Not Autoclavable | |
Cell culture, Molecular biology | |
1 Ea. | |
2.1 kg |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
epMotion 96 Flex tutorial Change of dispensing heads
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
9Hours
22Minutes
53Seconds
5
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ 96-channel Dispensing Heads > 0.5 to 300 μL
Spot an opportunity for improvement?Share a Content Correction