Learn More
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates
For high-throughput screening in cell based test systems.
347.00 € - 388.00 €
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Material | Polypropylene |
No. of Wells | 96 |
Well Shape | Round Bottom (U-well) |
Product Code | Brand | Color | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Sterility | Price | Quantity & Availability | ||||
10440912
|
Greiner Bio-One
650209 |
Black | Non-sterile |
388.00 €
Pack of 100 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
10779984
|
Greiner Bio-One
650261 |
Natural | Sterile |
350.00 €
Pack of 100 |
Estimated Shipment: 09-01-2025
Log in to see stock. |
||||
10304822
|
Greiner Bio-One
650205 |
Green | Non-sterile |
347.00 €
Pack of 100 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
13052133
|
Greiner Bio-One
650203 |
Red | Non-sterile |
347.00 €
Pack of 100 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
10433075
|
Greiner Bio-One
650207 |
White | Non-sterile |
388.00 €
Pack of 100 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
||||
13072133
|
Greiner Bio-One
650206 |
Yellow | Non-sterile |
347.00 €
Pack of 100 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
Description
- Non-pyrogenic
- Conical well design with flat bottom
- Ensures accurate pipetting
- Reduces dead volume of well
- Free of detectable DNase, RNase, human DNA
- Prevents wicking
Specifications
Standard | |
Polypropylene | |
Round Bottom (U-well) |
8 x 12 | |
96 |
Safety and Handling
missing translation for 'shelfLife' : 5 Years