Learn More
Greiner Bio-One 96-well CrystalQuick™ Crystallisation Microplates
Product Code. 10010372
Click to view available options
Well Shape:
Round
Square, Flat Bottom (F-well)
Volume (Metric) Well:
1.9 μL
3.9 μL
4.1 μL
Product Type:
LBR
LBR (low birefringence)
Standard
Unit Size:
Pack of 40
Pack of 80
Product Code. 10010372
Brand: Greiner Bio-One 609101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.
  • Non-sterile
  • Polystyrene
  • Good optical properties
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, experiments are possible with sitting drop and hanging drop metho
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color Clear
Product Type Standard
Material Polystyrene
Volume (Metric) Well 4.1 μL
No. of Wells 96
Sterility Non-sterile
Well Shape Square, Flat Bottom (F-well)
For Use With (Application) Sitting drop applications with different well profiles and material properties
No. per Case 40
Quantity 40/Cs.
Height (Metric) 14.4 mm