Learn More
Fisherbrand™ 96-Well Microplate Holder
Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W x H):
54 x 19 x 19 mm
128 x 86 x 23 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | 96-well Microplate Holder |
| Dimensions (L x W x H) | 54 x 19 x 19 mm |
| For Use With (Equipment) | Fisherbrand™ Microplate Shaker and Incubating Microplate Shaker |
| Includes | 7 pieces per pack |