missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AAK1, GST Tag Recombinant Protein
This AAK1 Recombinant Human Protein (1-465) was expressed in insect cells.
524.00 € - 7580.00 €
Specifications
Accession Number | AAH02695 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 77.3 kDa |
KinaseFamily | NAK Kinase Family |
Protein Length | aa 1 - 510 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970293
|
Invitrogen™
A30967 |
10 μg |
524.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15950383
|
Invitrogen™
A32880 |
100 μg |
2435.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17168785
|
Invitrogen™
A33370 |
1 mg |
7580.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AAK1 serine/threonine kinase is a member of the NAK family and plays a regulatory role in endocytosis. The purified catalytic domain (1-510) of AAK1 can be used in a number of different applications, but has been specifically validated for use with our LanthaScreen™ Kinase binding assay technology to identify novel kinase inhibitors.- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Specifications
AAH02695 | |
77.3 kDa | |
aa 1 - 510 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Baculovirus | |
Recombinant, Catalytic |
Kinase Assay | |
NAK Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |