Learn More
Krüss Optronic™ Abbe AR4 Analogue Refractometer
Brand: Krüss Optronic™ AR4
Specifications
Analog Refractometer | |
±0.0002nD | |
110/220 V |
1.3 to 1.7 | |
ASTM D1218 | |
110/220 V |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.