Learn More
Abnova™ TOP2A/CEN17q (Orange/Green) FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0119.200uL
Product Code. 16093175
  • 2495.00 € / 200µL
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
Store at -20°C in the dark.
Orange, Green
17q21/17p11.1-17q11.1
TOP2A/CEN17q FISH Probe
Genomic DNA
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Fluorescent In Situ Hybridization
Approx. 166kb/1.02kb
200 μL