missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Human Abundant Serum Markers Panel, 26plex

The ProcartaPlex Human Abundant Serum Markers Panel 26plex enables the study of immune function by analyzing 26 protein markers in a single well using Luminex xMAP technology.

Brand:  Invitrogen™ EPX260-15808-901

Product Code. 15994264

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The ProcartaPlex Human Abundant Serum Markers Panel 26plex enables the study of immune function by analyzing 26 protein markers in a single well using Luminex xMAP technology. The protein targets provided in this panel appear to be highly abundant in serum and plasma. Therefore, simultaneous detection of these targets may lead to efficient immune response profiling, biomarker discovery, and validation. The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup. It is not combinable with simplexes or other panels.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

Target list [bead region]: Adiponectin [57], CD14 [48], CD44 [43], CD44var (v6) [22], CD62L (L-Selectin) [54], Cystatin C [34], Elafin [64], Endoglin [46], ICAM-1 [73], Lactoferrin [77], Leptin [45], MBL [55], MMP-3 [19], MMP-9 [20], NGAL [14], NRP-1 [65], Osteopontin (OPN) [67], PAI-1 (Serpin) [35], RANTES (CCL5) [42], RBP4 [33], REG-3a [53], Resistin (ADSF) [39], SCGF beta [36], TIMP-1 [26], VCAM-1 [74], ZAG [47]

Recommended sample pre-dilution: Serum, Plasma: 1:100

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Multiplex Panel
Human
Fluorescence
See Certificate of Analysis
Luminex™ Instruments
Serum, Plasma, Cell Culture Supernatants

• 2 vials Human Standard Mix N1 (lyophilized)
• 1 vial Capture Bead Mix (1X)
• 1 vial Biotinylated Detection Antibody Mix (1X)
• 1 Bottle Reading Buffer (1X)
• 1 Bottle Wash Buffer (10X)
• 1 Bottle Streptavidin-PE (1X)
• 1 Bottle Universal Assay Buffer (10X)
• 8-tube strip
• Adhesive film
• Flat-bottom 96-well plate, black

Store at 2°C to 8°C

Immunology
96 Tests
Non-Combinable
See Certificate of Analysis
Multiplex Kit
ProcartaPlex™
Serum, Plasma: 25 μL, CCS: 50 μL
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.