Learn More
BINDER™ Access Port with Silicone Plug
Product Code. 15975052 Shop All BINDER Products
Click to view available options
Diameter (Metric):
100 mm
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 15975052
Brand: BINDER™ 80121671
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Access port with silicone plug for drying and heating chambers
  • Top Access Port is an accessory for use on the top of chambers and incubators
  • Silicone plug
  • 100 mm diameter
Specifications
Description Access port with silicone plug - 100 mm, top
For Use With (Equipment) BINDER Chambers FD 56, FD 115, FD 260, FD 720
Type Access Port
Includes Silicone Plug
Diameter (Metric) 100 mm