Learn More
BINDER™ Access Port with Silicone Plug
Shop All BINDER Products
Click to view available options
Diameter (Metric):
100 mm
Unit Size:
Each
Description
- Top Access Port is an accessory for use on the top of chambers and incubators
- Silicone plug
- 100 mm diameter
Specifications
Specifications
| Description | Access port with silicone plug - 100 mm, top |
| For Use With (Equipment) | BINDER Chambers FD 56, FD 115, FD 260, FD 720 |
| Type | Access Port |
| Includes | Silicone Plug |
| Diameter (Metric) | 100 mm |