Learn More
Axygen™ Accessories for Horizontal Gel Box, 10-20 cm
Product Code. p-7075336 Shop All Axygen Scientific Products
Click to view available options
No. of Wells:
10
12
16
20
25
30
40
50
8
Not Applicable
Type:
Casting Dam
Comb
Multi-channel Comb
Replacement Negative Electrode
Replacement Positive Electrode
UV Tray
Size:
10 cm
20 cm
10 x 10 cm
20 x 20 cm
Thickness (Metric):
0.75 mm
1 mm
1.5 mm
2 mm
Not Applicable
Unit Size:
1 unit
Product Code. 15593097
Brand: Axygen HGB1010MC1
Only null left
Add to basket
Add to basket
For use with horizontal gel box, 10cm.
Combs for 10-20 cm Axygen™ horizontal gel box
TRUSTED_SUSTAINABILITY
Specifications
Description Horizontal Electrophoresis System Accessories
For Use With (Equipment) Axygen Horizontal Gel Box, 10 cm
Type Multi-channel Comb
No. of Wells 10
Size 10 cm
Thickness (Metric) 1 mm