Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Digital Heating Shaking Drybath Product Code.: 15238884
Add to basket
Thermo Scientific™ Accessories for Thermo Scientific™ Digital Heating Shaking Drybath
Product Code. 15238884 Shop All Thermo Scientific Products
1.5mL
Click to view available options
Tube Size:
1.5mL
Product Code. 15238884
Brand: Thermo Scientific™ 88880127
Edge
Add to basket
Add to basket
Optimize flexibility and performance
TRUSTED_SUSTAINABILITY
Specifications
No. of Places 24
Type Accessory, Block Set with Cover
Tube Size 1.5mL
  • Multiple Manufacturers
    Up to 60% off
    Promo Code24119
    Expires2025-04-30
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Days
    43
    Hours
    15
    Minutes
    30
    Seconds
    10
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Accessories for Thermo Scientific™ Digital Heating Shaking Drybath >