Learn More
Hamilton™ AccuDil™ Syringe
Click to view available options
Volume (Metric):
2.5 mL
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inner | 7.2 mm |
| Diameter (Metric) Outer | 9.6 mm |
| Product Type | AccuDil Syringe |
| Max. Pressure | 200 psi |
| Model | 1002 |
| Quantity | 1/Ea. |
| Volume (Metric) | 2.5 mL |
| For Use With (Equipment) | ML500 Series Diluters and Dispensers |
| Connection Type | Special |