Learn More
Key Instruments™ Flo-Rite™ Acrylic Airflow Flowmeter
Click to view available options
Airflow Rate:
0.04 to 0.5 L/min.
0.1 to 1 L/min.
1 L/min.
1 to 10 L/min.
5 L/min.
Height (Metric):
102 mm
165 mm
Unit Size:
Each
Description
FEATURES
- Easy-to-read English or Metric
- Scales Water ranges from 4 CCM to 20 GPM
- Air ranges from 40 CCM to 4000 LPM
- Threaded brass inserts for quick installation
- Easy disassembly and assembly for maintenance
- Durable one-piece clear acrylic construction
- Stable, easy-to-read float Superior quality
- Air sampling equipment aquaculture
- Desalinization equipment
- Gas analyzers Medical systems
- Photoprocessing equipment
- Water treatment and distribution systems
Specifications
Specifications
| Type | Flowmeter |
| Accuracy | >±5% |
| Airflow Rate | 0.04 to 0.5 L/min. |
| Pressure Rating | 100 psig |
| For Use With (Application) | Measuring Liquid or Gas Flows in Air Sampling Equipment, Chromatography Systems, Desalinization Equipment, Gas Analyzers, Medical Systems, Photo Equipment, Water Treatment and Distribution and in Aquaculture System |
| Height (Metric) | 102 mm |
| Internal Part Material | Acrylic |
| Repeatability | ±0.5% |
| Width (Metric) | 26 mm |
| Scale | 50, 100 or 127 mm |
| Show More |