Learn More
Macherey-Nagel™ Activity Mixture
Brand:  Macherey-Nagel™ 722307
Product Code. 10717282
  • 78.35 € / 1mL
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Specifications
For GC Capillary Columns
1 mg/mL each of TMS Capric Acid (C10), TMS Myristic Acid (C14), TMS Stearic Acid (C18), TMS Behenic Acid (C22), Hexadecane (C16), Eicosane (C20), Tetracosane (C24), Octacosane (C28)
1 mL
Activity Mixture
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Activity Mixture > Capacity: 1mL