missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA584258
This item is not returnable.
View return policy
Description
Immunogen sequence: IIPQWKTSES PVREKHPLKA ELRVMAEGRE LILDLEKNEQ LFAPSYTETH YTSSGNPQTT TRKLEDHCFY HGTVRETELS SVTLSTCRGI RGLITVSSNL SYVIEPLPDS KGQHLIYRSE HLKPPPGNCG FEHSKP
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a type I transmembrane protein and serves as a marker for dendritic cell differentiation. It has also been demonstrated to be an active metalloproteinase, which may be involved in normal physiological and pathological processes such as cells migration, cell adhesion, cell-cell and cell-matrix interactions, and signal transduction.
Specifications
| ADAM19 | |
| Polyclonal | |
| Unconjugated | |
| ADAM19 | |
| a disintegrin and metallopeptidase domain 19 (meltrin beta); a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 19 (meltrin beta); ADAM; ADAM 19; ADAM metallopeptidase domain 19; ADAM19; ADAMs; AL024287; disintegrin and metalloproteinase domain-containing protein 19; FKSG34; MADDAM; meltrin beta; meltrin-beta; metalloendopeptidases; Metalloprotease and disintegrin dendritic antigen marker; metalloprotease-disintegrin meltrin beta; MLTNB; Sox30; SRY-box containing gene 30 | |
| Rabbit | |
| Anitgen affinity chromatography, Antigen affinity chromatography | |
| RUO | |
| 8728 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H013 | |
| ADAM19 | |
| Recombinant protein corresponding to Human ADAM19 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction