Learn More
Memmert™ Additional Stainless Steel Grid for UF1060, Reinforced Chamber Option
Product Code. 17538805 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF1060 Oven
Unit Size:
Each
Product Code. 17538805
Brand: Memmert™ UF1060B32550
Only null left
Add to basket
Add to basket
Additional grid features a sturdy stainless steel construction.
  • Reinforced
  • Stainless Steel
  • Electropoloished
  • Max. loading 60 kg
Specifications
For Use With (Equipment) UF1060 Oven
Type Reinforced Grid
Material Stainless Steel
Max. Shelf Load 60 kg