Learn More
Memmert™ Additional Stainless Steel Grid for UN160plus, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Brand:  Memmert™ UN160plusE29767
Product Code. 17589915
  • 298.00 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reinforced
  • Stainless Steel
  • Electropoloished
  • Max. loading 60 kg
Specifications
Specifications
UN160plus Oven
Reinforced Grid
Stainless Steel
60 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Additional Stainless Steel Grid for UN160plus, Reinforced Chamber Option > For Use With: UN160plus