Learn More
Brady™ Adhesive Polyester Prohibition Signs
Brand: Brady™ 251402
This item is not returnable.
View return policy
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC | |
Polyester | |
Pipe markers and safety signs | |
297 mm | |
Access forbidden to Unauthorized Persons |
STN 652-297X105-B7541 (BRANDED) | |
Adhesive Polyester | |
105 mm | |
Rectangular | |
Prohibition Sign |