Learn More
B Braun™ Aesculap™ Delicate Iris Scissors for Plastic Surgery
Small scissors with fine and sharp points for delicate work in applications such as plastic surgery.
Brand: B Braun™ BC056R
This item is not returnable.
View return policy
Description
- Manufactured from stainless steel
- Meticulously crafted to have a solid feel and to perform like an extension of the surgeon’s own hand
- Manufactured to strict guidelines for precision and reliability
- Round handles
- Available with straight or curved blades
Specifications
Iris Scissors | |
95 mm | |
Delicate surgical applications |
Straight | |
Stainless Steel | |
Sharp/Sharp |