Learn More
B Braun™ Aesculap™ Safety Scalpel
Reduces the risk of infections caused by cuts and stab wounds suffered during handover, take-over and disposal of scalpels.
Brand:  B Braun™ BA815SU
 View more versions of this product
Product Code. 15772577
  • 43.00 € / Pack of 10
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Safety locking mechanism for disposal
  • Single-handed operation
  • Ergonomic guide for precise incision
  • Ruler in cm and inches
  • Transparent cap for blade protection – blade and position is always visible
  • Provides optimal sharpness
Specifications
Specifications
Carbon Steel
Surgeon Grade
Sterile
Plastic
# 15
Single-use
Protective Cap
10/Pk.