Learn More
Invitrogen™ AFP Monoclonal Antibody (P5B8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA514666
Description
MA5-14666 can be used for immunofluorescence analysis of AFP in the endoderm derived from human embryonic stem cells. By Western blot, MA5-14666 detects endogenous AFP protein in the early hepatocyte-like cells derived from human embryonic stem cells. Product MA514666 is a smaller package size of MIA1305 (formerly sold as a Seradyn product).
AFP (Alpha Fetoprotein) is a major plasma protein produced by the yolk sac, intestinal tract and the liver during fetal life. AFP expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of AFP may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and AFP and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. AFP is found in monomeric, dimeric, and trimeric forms. AFP can also bind to copper ions, nickel ions, fatty acids and bilirubin. The level of AFP in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. High level of serum AFP has been identified in patients with hepatocellular carcinomas (HCC), teratoblastoma, colorectal cancer, pancreatic cancer, and germ cell neoplasms.
Specifications
| AFP | |
| Monoclonal | |
| 1 mg/mL | |
| 0.01M potassium phosphate with 1 mg/mL BSA, 0.135M NaCl, 2.5mM KCl and 0.05% sodium azide | |
| P02771 | |
| AFP | |
| Purified AFP from Cord Blood. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry | |
| P5B8 | |
| Unconjugated | |
| AFP | |
| a fetoglobulin; a fetoprotein; Afp; AFPD; Alpha fetoglobulin; alpha fetoprotein; alpha-1-fetoprotein; Alpha-fetoglobulin; alpha-fetoprotein; alpha-fetoprotein precursor; alpha-foetoprotein; chloride intracellular channel 6; FETA; fetoglobulin; fetoprotein; HPAFP; α fetoglobulin; α fetoprotein | |
| Mouse | |
| Protein A | |
| RUO | |
| 174 | |
| Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.