missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Isoelectric Focusing of Proteins/IEF Grade), Fisher BioReagents

Product Code. 11818612
Click to view available options
Poly Bottle; 25g
This item is not returnable. View return policy
This item is not returnable. View return policy
Highly purified agarose for separating proteins by isoelectric focusing.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Powder/Solid
CAS 9012-36-6
Color Cream
Quantity 25 g
DNase DNase free
Electroendosmosis (EEO) Not Detected
Molecular Formula C12H18O9
Gelation Temperature 34° to 45°C
Gel Strength 500g/cm2min.
Formula Weight 306.12
Grade EIF
Packaging Poly Bottle
Chemical Name or Material Agarose, Isoelectric Focusing of Proteins
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'